Assay Details
| Gene Symbol: Atp6v1e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007510.2 | NP_031536.2 | 7 - 8 | 593 | 78 | |
| GenBank mRNA | AB074758.1 | - | 7 - 8 | 595 | 78 | |
| AK010603.1 | - | 7 - 8 | 609 | 78 | ||
| AK029146.1 | - | 1380 | 78 | |||
| AK139791.1 | - | 7 - 8 | 610 | 78 | ||
| AK146162.1 | - | 7 - 8 | 607 | 78 | ||
| AK149356.1 | - | 7 - 8 | 629 | 78 | ||
| AK167644.1 | - | 7 - 8 | 552 | 78 | ||
| AK169854.1 | - | 7 - 8 | 607 | 78 | ||
| BC003421.1 | - | 7 - 8 | 593 | 2598896 | 78 | |
| BC055438.1 | - | 7 - 8 | 627 | 3995744 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atp6v1e1
Genomic Map