Assay Details
| Gene Symbol: Ccdc77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026028.5 | NP_080304.1 | 1 - 2 | 141 | 96 | |
| GenBank mRNA | CD555913.1 | - | 1 - 2 | 141 | 96 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_026028.5 transcripts.
Set Membership: |
|

Ccdc77
Genomic Map