Assay Details
Gene Symbol: Renbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164704.1 | NP_001158176.1 | 4 - 5 | 353 | 59 | |
NM_023132.3 | NP_075621.3 | 4 - 5 | 395 | 59 | ||
GenBank mRNA | AK014347.1 | - | 4 - 5 | 315 | 59 | |
AK146652.1 | - | 4 - 5 | 360 | 59 | ||
AK167648.1 | - | 4 - 5 | 266 | 59 | ||
AY183138.1 | - | 4 - 5 | 363 | 59 | ||
BC014821.1 | - | 4 - 5 | 289 | 4238244 | 59 | |
BC035455.1 | - | 4 - 5 | 289 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
