Assay Details
Gene Symbol: Renbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164704.1 | NP_001158176.1 | 5 - 6 | 533 | 89 | |
NM_023132.3 | NP_075621.3 | 5 - 6 | 575 | 89 | ||
GenBank mRNA | AK014347.1 | - | 5 - 6 | 495 | 89 | |
AK146652.1 | - | 5 - 6 | 540 | 89 | ||
AK167648.1 | - | 5 - 6 | 446 | 89 | ||
AK186123.1 | - | 1 - 2 | 81 | 89 | ||
AY183138.1 | - | 5 - 6 | 543 | 89 | ||
BC014821.1 | - | 5 - 6 | 469 | 4238244 | 89 | |
BC035455.1 | - | 5 - 6 | 469 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
