Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600021235.mm.1 |
TC0600002728.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600021235.mm.1 |
NM_133928.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021235.mm.1 |
NM_133928.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021236.mm.1 |
TC0600002728.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600021236.mm.1 |
NM_133928.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021236.mm.1 |
NM_133928.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021237.mm.1 |
TC0600002728.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600021237.mm.1 |
NM_133928.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021237.mm.1 |
NM_133928.2 |
Refseq |