Assay Details
| Gene Symbol: Hdac11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144919.2 | NP_659168.1 | 1 - 2 | 33 | 74 | |
| GenBank mRNA | AK045994.1 | - | 1 - 2 | 45 | 74 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK045994.1, NM_144919.2 transcripts.
Set Membership: |
|

Hdac11
Genomic Map