Assay Details
| Gene Symbol: Gmcl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011818.3 | NP_035948.3 | 2 - 3 | 592 | 148 | |
| GenBank mRNA | AF163665.1 | - | 2 - 3 | 536 | 148 | |
| AF186095.1 | - | 2 - 3 | 428 | 148 | ||
| AF282322.1 | - | 2 - 3 | 462 | 148 | ||
| AK004716.1 | - | 2 - 3 | 568 | 148 | ||
| AK132788.1 | - | 2 - 3 | 456 | 148 | ||
| AK146964.1 | - | 2 - 3 | 561 | 148 | ||
| AK155238.1 | - | 2 - 3 | 575 | 148 | ||
| BC054767.1 | - | 2 - 3 | 551 | 6414943 | 148 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gmcl1
Genomic Map