Assay Details
| Gene Symbol: Gmcl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011818.3 | NP_035948.3 | 4 - 5 | 791 | 59 | |
| GenBank mRNA | AF163665.1 | - | 4 - 5 | 735 | 59 | |
| AF186095.1 | - | 4 - 5 | 627 | 59 | ||
| AK004716.1 | - | 4 - 5 | 767 | 59 | ||
| AK146964.1 | - | 4 - 5 | 760 | 59 | ||
| AK155238.1 | - | 4 - 5 | 774 | 59 | ||
| BC054767.1 | - | 4 - 5 | 750 | 6414943 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gmcl1
Genomic Map