Assay Details
| Gene Symbol: Snrpg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026506.2 | NP_080782.1 | 3 - 4 | 226 | 71 | |
| GenBank mRNA | AK011129.1 | - | 3 - 4 | 225 | 71 | |
| AK012802.1 | - | 3 - 4 | 215 | 71 | ||
| AK013235.1 | - | 3 - 4 | 227 | 71 | ||
| AK017852.1 | - | 3 - 4 | 575 | 71 | ||
| BC027499.1 | - | 3 - 4 | 199 | 1430048 | 71 | |
| BC051470.1 | - | 3 - 4 | 271 | 1244608 | 71 | |
| BC094411.1 | - | 3 - 4 | 213 | 1380524 | 71 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24609764 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24609767 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24609765 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24609766 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1990081 | BC094411.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1990081 | AK017852.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1990081 | BC027499.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1990081 | BC051470.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1990081 | NM_026506.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2007303 | BC094411.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2007303 | AK017852.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2007303 | BC027499.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2007303 | BC051470.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2007303 | NM_026506.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 896413 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 621521 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 144928 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 337236 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 75533 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P175731 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2007303 | BC094411.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2007303 | AK017852.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2007303 | BC027499.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2007303 | BC051470.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2007303 | NM_026506.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1990081 | BC094411.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1990081 | AK017852.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1990081 | BC027499.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1990081 | BC051470.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1990081 | NM_026506.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P200134 | BC094411.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P200134 | AK017852.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P200134 | BC027499.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P200134 | BC051470.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P200134 | NM_026506.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24609761 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24609780 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24609766 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24609771 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24609763 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24609772 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24609775 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24609770 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24609762 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24609764 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24609767 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24609777 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24609774 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24609757 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24609779 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24609768 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24609778 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24609759 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P175731 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05968756 | - | - |
| Whole Mouse Genome G4122A | A_51_P200134 | BC094411.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P200134 | AK017852.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P200134 | BC027499.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P200134 | BC051470.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P200134 | NM_026506.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121180 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103540 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P175731 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05335296 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05513578 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05884243 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05513574 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05513576 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05513573 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24609764 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24609767 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24609765 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24609766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609713 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05884242 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609774 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05468021 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609780 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05513574 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05054659 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609772 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05513578 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05679678 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609762 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05513573 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609797 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609784 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05679680 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05513576 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05679679 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609770 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609793 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609786 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05727982 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609800 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609775 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05780491 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609788 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609783 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609804 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609781 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609759 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609805 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609760 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05098058 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609757 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609778 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10539706 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539708 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539709 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539707 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449244 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448357_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10449242 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10539705 | - | - |
| Mouse Genome 430 2.0 Array | 1448358_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1448358_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1448357_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448357_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600007606.mm.1 | TC0600000986.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600007606.mm.1 | NM_026506.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600007606.mm.1 | NM_026506.2 | Refseq |
| Mouse Expression Set 430 | 1448357_at | - | - |
| Mouse Expression Set 430 | 1448358_s_at | - | - |
| Mouse Expression Set 430 | 1448358_PM_s_at | - | - |
| Mouse Expression Set 430 | 1448357_PM_at | - | - |
| Murine Genome U74v2 Set | 93999_PM_at | - | - |
| Murine Genome U74v2 Set | 93999_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448357_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448358_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448357_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448358_PM_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 647289 | - | - |
| ABI Mouse Genome Survey Microarray | 895099 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2340551 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 104010133 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2340551 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002340551 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2442140 | - | - |
| mouse-6 v1.1 (Illumina) | scl068011.3_46-S | - | - |
| mouse-6 v1.1 (Illumina) | scl29806.1_289-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 104010133 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2340551 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl068011.3_46-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2340551 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 104010133 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2975640 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2742647 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2975640 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2442140 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl068011.3_46-S | - | - |

Snrpg
Genomic Map