Assay Details
| Gene Symbol: Aak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040106.2 | NP_001035195.1 | 8 - 9 | 1508 | 105 | |
| NM_177762.6 | NP_808430.2 | 8 - 9 | 1508 | 105 | ||
| GenBank mRNA | AK147363.1 | - | 8 - 9 | 1509 | 105 | |
| AK158879.1 | - | 8 - 9 | 1398 | 105 | ||
| BC141176.1 | - | 8 - 9 | 1264 | 9055788 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aak1
Genomic Map