Assay Details
| Gene Symbol: Sec61a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016906.4 | NP_058602.1 | 7 - 8 | 719 | 57 | |
| GenBank mRNA | AB032902.1 | - | 7 - 8 | 731 | 57 | |
| AF145253.1 | - | 7 - 8 | 641 | 57 | ||
| AF222743.1 | - | 7 - 8 | 641 | 57 | ||
| AK054318.1 | - | 2 - 3 | 1555 | 57 | ||
| AK088473.1 | - | 7 - 8 | 720 | 57 | ||
| AK153055.1 | - | 7 - 8 | 709 | 57 | ||
| AK159349.1 | - | 7 - 8 | 707 | 57 | ||
| AK167484.1 | - | 7 - 8 | 649 | 57 | ||
| AK197110.1 | - | 3 - 4 | 331 | 57 | ||
| BC003707.1 | - | 7 - 8 | 645 | 3592772 | 57 | |
| BC006020.1 | - | 3 - 4 | 304 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sec61a1
Genomic Map