Assay Details
| Gene Symbol: Sec61a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016906.4 | NP_058602.1 | 9 - 10 | 1086 | 104 | |
| GenBank mRNA | AB032902.1 | - | 9 - 10 | 1098 | 104 | |
| AF145253.1 | - | 9 - 10 | 1008 | 104 | ||
| AF222743.1 | - | 9 - 10 | 1008 | 104 | ||
| AK054318.1 | - | 4 - 5 | 1922 | 104 | ||
| AK088473.1 | - | 9 - 10 | 1087 | 104 | ||
| AK153055.1 | - | 9 - 10 | 1076 | 104 | ||
| AK159349.1 | - | 9 - 10 | 1074 | 104 | ||
| AK167484.1 | - | 9 - 10 | 1016 | 104 | ||
| AK206477.1 | - | 1 - 2 | 187 | 104 | ||
| BC003707.1 | - | 9 - 10 | 1012 | 3592772 | 104 | |
| BC006020.1 | - | 5 - 6 | 671 | 104 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sec61a1
Genomic Map