Assay Details
| Gene Symbol: Mgll | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166249.1 | NP_001159721.1 | 1 - 2 | 163 | 134 | |
| NM_011844.4 | NP_035974.1 | 1 - 2 | 163 | 134 | ||
| GenBank mRNA | AK028008.1 | - | 1 - 2 | 106 | 134 | |
| AK131645.1 | - | 1 - 2 | 143 | 134 | ||
| AK148242.1 | - | 1 - 2 | 106 | 134 | ||
| AK148395.1 | - | 1 - 2 | 106 | 134 | ||
| BC057965.1 | - | 1 - 2 | 88 | 4986023 | 134 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK131645.1, AK148242.1, AK148395.1, BC057965.1, NM_001166249.1, NM_011844.4 transcripts.
Set Membership: |
|

Mgll
Genomic Map