Assay Details
| Gene Symbol: Rpn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133933.4 | NP_598694.3 | 2 - 3 | 386 | 87 | |
| GenBank mRNA | AK029923.1 | - | 2 - 3 | 344 | 87 | |
| AK052130.1 | - | 2 - 3 | 340 | 87 | ||
| AK132750.1 | - | 2 - 3 | 359 | 87 | ||
| AK151899.1 | - | 2 - 3 | 350 | 87 | ||
| AK152003.1 | - | 2 - 3 | 359 | 87 | ||
| AK153867.1 | - | 2 - 3 | 345 | 87 | ||
| BC016080.1 | - | 2 - 3 | 348 | 4489287 | 87 | |
| BC085483.1 | - | 2 - 3 | 327 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rpn1
Genomic Map