Assay Details
| Gene Symbol: Tacstd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020047.3 | NP_064431.2 | 1 | 1714 | 103 | |
| GenBank mRNA | AK077370.1 | - | 1 | 1689 | 103 | |
| AK085753.1 | - | 1 | 1689 | 103 | ||
| AK155470.1 | - | 1 | 987 | 103 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of epithelial cell migration
negative regulation of stress fiber assembly
negative regulation of branching involved in ureteric bud morphogenesis
negative regulation of substrate adhesion-dependent cell spreading
negative regulation of ruffle assembly
negative regulation of cell motility
positive regulation of stem cell differentiation
molecular_function
negative regulation of epithelial cell migration
negative regulation of stress fiber assembly
negative regulation of branching involved in ureteric bud morphogenesis
negative regulation of substrate adhesion-dependent cell spreading
negative regulation of ruffle assembly
negative regulation of cell motility
positive regulation of stem cell differentiation
molecular_function
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24521962 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24521961 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24521965 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24521958 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24521964 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24521957 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24521963 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24521959 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24521960 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P257938 | BC117719.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P257938 | BC117720.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P257938 | AK085753.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P257938 | NM_020047.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P257938 | AK076410.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P257938 | BC117719.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P257938 | BC117720.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P257938 | AK085753.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P257938 | NM_020047.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P257938 | AK076410.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 914934 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P164264 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P178017 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P257938 | BC117719.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P257938 | BC117720.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P257938 | AK085753.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P257938 | NM_020047.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P257938 | AK076410.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P257938 | BC117719.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P257938 | BC117720.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P257938 | AK085753.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P257938 | NM_020047.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P257938 | AK076410.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P257938 | BC117719.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P257938 | BC117720.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P257938 | AK085753.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P257938 | NM_020047.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P257938 | AK076410.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P257938 | BC117719.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P257938 | BC117720.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P257938 | AK085753.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P257938 | NM_020047.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P257938 | AK076410.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24521971 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24521946 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24521967 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24521981 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24521973 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24521962 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24521944 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24521957 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24521976 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24521965 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24521961 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24521977 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24521948 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24521972 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24521949 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24521951 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24521954 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24521978 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24521953 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24521963 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24521955 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24521986 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24521983 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24521988 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24521966 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24521969 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24521974 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24521989 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24521980 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24521958 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P164264 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P178017 | - | - |
| Whole Mouse Genome G4122A | A_51_P257938 | BC117719.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P257938 | BC117720.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P257938 | AK085753.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P257938 | NM_020047.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P257938 | AK076410.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P257938 | BC117719.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P257938 | BC117720.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P257938 | AK085753.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P257938 | NM_020047.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P257938 | AK076410.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100822 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P178017 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P164264 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05377545 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05377564 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05377559 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05132719 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05132727 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05377529 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24521964 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24521963 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24521957 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24521958 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24521962 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24521960 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24521959 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24521965 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24521961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24521969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24521951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24521937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24521938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24522003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24521911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24521978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24521988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24521919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24521928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24521983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24521926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24521935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24521963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24521986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24522008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24521973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24521976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24521958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24521977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24521972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24521981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24521934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24521954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24521967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24521909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24522000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24521923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24521946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24521924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24521989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24521908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24521906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24521949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24521916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24521941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24521922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24521914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24522011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24521930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24522006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24521915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24521966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24521980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24521961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24521944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24521974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24522001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24521948 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24521916 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24521951 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24521932 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24521954 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24521969 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05377542 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24521914 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24521986 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24521962 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05063572 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24521926 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05443505 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24521915 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24521974 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24521977 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24521928 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24521966 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24521958 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05377545 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24521983 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24521940 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05377557 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24521976 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24521989 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05377568 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24521924 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24521972 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24521935 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24521934 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24521944 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24521988 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05377570 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24521930 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24521980 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24521922 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24521963 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05377538 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24521942 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24521938 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24521973 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24521981 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24521923 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24521971 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24521946 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05122666 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24521949 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10545170 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10545172 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10545171 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10545169 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423323_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10545168 | - | - |
| Mouse Genome 430 2.0 Array | 1423323_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1423323_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56753_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56753_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600018629.mm.1 | TC0600002427.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600018629.mm.1 | NM_020047.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600018629.mm.1 | NM_020047.3 | Refseq |
| Mouse Expression Set 430 | 1423323_at | - | - |
| Mouse Expression Set 430 | 1423323_PM_at | - | - |
| Murine Genome U74v2 Set | 160651_PM_at | - | - |
| Murine Genome U74v2 Set | 103648_at | - | - |
| Murine Genome U74v2 Set | 103648_PM_at | - | - |
| Murine Genome U74v2 Set | 160651_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423323_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423323_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423323_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423323_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 329002 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6660341 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6660341 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006660341 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1231513 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1231513 | - | - |
| mouse-6 v1.1 (Illumina) | scl28929.1_28-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6660341 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl28929.1_28-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6660341 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1231513 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1231513 | - | - |

Tacstd2
Genomic Map