Assay Details
| Gene Symbol: Il12rb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008354.3 | NP_032380.1 | 3 - 4 | 539 | 92 | |
| GenBank mRNA | BC137745.1 | - | 2 - 3 | 451 | 8860764 | 92 |
| U64199.1 | - | 3 - 4 | 540 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Il12rb2
Genomic Map