Assay Details
Gene Symbol: Gkn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025466.1 | NP_079742.1 | 3 - 4 | 239 | 84 | |
GenBank mRNA | AK008622.1 | - | 3 - 4 | 239 | 84 | |
AK008640.1 | - | 3 - 4 | 237 | 84 | ||
AK008641.1 | - | 3 - 4 | 239 | 84 | ||
AK008647.1 | - | 3 - 4 | 236 | 84 | ||
AK008722.1 | - | 3 - 4 | 237 | 84 | ||
AK008745.1 | - | 3 - 4 | 239 | 84 | ||
AK008933.1 | - | 3 - 4 | 241 | 84 | ||
AK008956.1 | - | 3 - 4 | 236 | 84 | ||
AK008990.1 | - | 3 - 4 | 241 | 84 | ||
AK009145.1 | - | 3 - 4 | 237 | 84 | ||
AK019050.1 | - | 3 - 4 | 240 | 84 | ||
AK028136.1 | - | 3 - 4 | 235 | 84 | ||
AY456960.1 | - | 3 - 4 | 239 | 84 | ||
BC099446.1 | - | 3 - 4 | 222 | 30521253 | 84 | |
BC132540.1 | - | 3 - 4 | 229 | 40130817 | 84 | |
BC132542.1 | - | 3 - 4 | 229 | 40130819 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
