Assay Details
| Gene Symbol: Prokr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021381.3 | NP_067356.2 | 1 - 2 | 106 | 66 | |
| GenBank mRNA | AK134279.1 | - | 1 - 2 | 120 | 66 | |
| BC059003.1 | - | 1 - 2 | 106 | 6848525 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK134279.1, BC059003.1, NM_021381.3 transcripts.
Set Membership: |
|

Prokr1
Genomic Map