Assay Details
| Gene Symbol: Arhgap25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037727.2 | NP_001032816.1 | 5 - 6 | 1047 | 98 | |
| NM_001286610.1 | NP_001273539.1 | 4 - 5 | 847 | 98 | ||
| NM_175476.4 | NP_780685.2 | 5 - 6 | 718 | 98 | ||
| NR_104483.1 | 5 - 6 | 718 | 98 | |||
| GenBank mRNA | AK037710.1 | - | 5 - 6 | 780 | 98 | |
| AK137444.1 | - | 5 - 6 | 706 | 98 | ||
| AK137526.1 | - | 5 - 6 | 655 | 98 | ||
| AK159598.1 | - | 4 - 5 | 848 | 98 | ||
| AK159941.1 | - | 4 - 5 | 848 | 98 | ||
| AK169897.1 | - | 5 - 6 | 1024 | 98 | ||
| AK172878.1 | - | 3 - 4 | 377 | 98 | ||
| BC108400.1 | - | 2 - 3 | 328 | 98 | ||
| BC138751.1 | - | 4 - 5 | 626 | 8861773 | 98 | |
| BC138752.1 | - | 4 - 5 | 626 | 8861774 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap25
Genomic Map