Assay Details
| Gene Symbol: Arhgap25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037727.2 | NP_001032816.1 | 8 - 9 | 1374 | 89 | |
| NM_001286610.1 | NP_001273539.1 | 7 - 8 | 1174 | 89 | ||
| NM_175476.4 | NP_780685.2 | 8 - 9 | 1045 | 89 | ||
| GenBank mRNA | AK037710.1 | - | 8 - 9 | 1107 | 89 | |
| AK048450.1 | - | 8 - 9 | 1005 | 89 | ||
| AK137444.1 | - | 8 - 9 | 1033 | 89 | ||
| AK159598.1 | - | 7 - 8 | 1175 | 89 | ||
| AK159941.1 | - | 7 - 8 | 1175 | 89 | ||
| AK172878.1 | - | 6 - 7 | 704 | 89 | ||
| BC108400.1 | - | 5 - 6 | 655 | 89 | ||
| BC138751.1 | - | 7 - 8 | 953 | 8861773 | 89 | |
| BC138752.1 | - | 7 - 8 | 953 | 8861774 | 89 | |
More Information
Set Membership: |
|

Arhgap25
Genomic Map