Assay Details
| Gene Symbol: Isy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133934.4 | NP_598695.1 | 6 - 7 | 407 | 84 | |
| GenBank mRNA | AK173116.1 | - | 375 | 84 | ||
| BC020103.1 | - | 6 - 7 | 371 | 3673446 | 84 | |
| BC037695.1 | - | 6 - 7 | 4009615 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Isy1
Genomic Map