Assay Details
| Gene Symbol: Rpia | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009075.2 | NP_033101.2 | 2 - 3 | 353 | 84 | |
| GenBank mRNA | AK137235.1 | - | 2 - 3 | 188 | 84 | |
| BC053526.1 | - | 2 - 3 | 176 | 84 | ||
| L35034.1 | - | 2 - 3 | 333 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rpia
Genomic Map