Assay Details
| Gene Symbol: Aicda | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009645.2 | NP_033775.1 | 3 - 4 | 513 | 159 | |
| GenBank mRNA | AF132979.1 | - | 3 - 4 | 512 | 159 | |
| AK080144.1 | - | 2 - 3 | 512 | 159 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aicda
Genomic Map