Assay Details
| Gene Symbol: Iqsec3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033354.3 | NP_001028526.1 | 5 - 6 | 2281 | 85 | |
| NM_001289685.1 | NP_001276614.1 | 4 - 5 | 2010 | 85 | ||
| NR_110358.1 | 4 - 5 | 2010 | 85 | |||
| GenBank mRNA | AK122446.1 | - | 4 - 5 | 1997 | 85 | |
| AK147351.1 | - | 4 - 5 | 1956 | 85 | ||
| AK169437.1 | - | 5 - 6 | 2281 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Iqsec3
Genomic Map