Assay Details
Gene Symbol: Raf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029780.3 | NP_084056.1 | 1 - 2 | 397 | 101 | |
GenBank mRNA | AK036317.1 | - | 1 - 2 | 284 | 101 | |
AK141745.1 | - | 1 - 2 | 398 | 101 | ||
BC015273.1 | - | 1 - 2 | 276 | 4222659 | 101 | |
BC056439.1 | - | 1 - 2 | 316 | 101 | ||
BC092040.1 | - | 1 - 2 | 262 | 2645313 | 101 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK141745.1, BC015273.1, BC092040.1, NM_029780.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
