Assay Details
| Gene Symbol: Raf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029780.3 | NP_084056.1 | 3 - 4 | 745 | 64 | |
| GenBank mRNA | AB057655.1 | - | 2 - 3 | 290 | 64 | |
| AB057663.2 | - | 3 - 4 | 515 | 64 | ||
| AK036317.1 | - | 3 - 4 | 632 | 64 | ||
| AK141745.1 | - | 3 - 4 | 746 | 64 | ||
| BC015273.1 | - | 3 - 4 | 624 | 4222659 | 64 | |
| BC056439.1 | - | 3 - 4 | 664 | 64 | ||
| BC092040.1 | - | 3 - 4 | 610 | 2645313 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Raf1
Genomic Map