Assay Details
| Gene Symbol: Raf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029780.3 | NP_084056.1 | 6 - 7 | 1104 | 88 | |
| GenBank mRNA | AB057655.1 | - | 5 - 6 | 649 | 88 | |
| AB057663.2 | - | 6 - 7 | 874 | 88 | ||
| AK036317.1 | - | 6 - 7 | 991 | 88 | ||
| AK141745.1 | - | 6 - 7 | 1105 | 88 | ||
| BC015273.1 | - | 6 - 7 | 983 | 4222659 | 88 | |
| BC056439.1 | - | 6 - 7 | 1023 | 88 | ||
| BC092040.1 | - | 6 - 7 | 969 | 2645313 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Raf1
Genomic Map