Assay Details
| Gene Symbol: Foxm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008021.4 | NP_032047.4 | 5 - 6 | 1143 | 69 | |
| GenBank mRNA | AK049177.1 | - | 5 - 6 | 1084 | 69 | |
| BC065067.1 | - | 5 - 6 | 1143 | 6417437 | 69 | |
| BC075723.1 | - | 5 - 6 | 1162 | 6515008 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Foxm1
Genomic Map