Assay Details
| Gene Symbol: Ogg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010957.4 | NP_035087.3 | 2 - 3 | 631 | 67 | |
| GenBank mRNA | AF000669.1 | - | 2 - 3 | 509 | 67 | |
| AF003596.1 | - | 2 - 3 | 596 | 67 | ||
| AK146870.1 | - | 2 - 3 | 631 | 67 | ||
| AK165875.1 | - | 2 - 3 | 583 | 67 | ||
| BC138733.1 | - | 2 - 3 | 587 | 8861755 | 67 | |
| BC138736.1 | - | 2 - 3 | 587 | 8861758 | 67 | |
| U88621.1 | - | 2 - 3 | 599 | 67 | ||
| U96711.1 | - | 2 - 3 | 596 | 67 | ||
| Y11247.1 | - | 2 - 3 | 562 | 67 | ||
| Y13479.1 | - | 2 - 3 | 632 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ogg1
Genomic Map