Assay Details
| Gene Symbol: Atp2b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036684.2 | NP_001031761.1 | 17 - 18 | 3432 | 63 | |
| NM_009723.5 | NP_033853.1 | 19 - 20 | 3621 | 63 | ||
| GenBank mRNA | AB030737.1 | - | 16 - 17 | 2993 | 63 | |
| AB030738.1 | - | 16 - 17 | 2993 | 63 | ||
| AF053471.1 | - | 18 - 19 | 3450 | 63 | ||
| AK054112.1 | - | 21 - 22 | 3719 | 63 | ||
| AK147240.1 | - | 19 - 20 | 3507 | 63 | ||
| AK147357.1 | - | 17 - 18 | 3361 | 63 | ||
| AK147369.1 | - | 19 - 20 | 3556 | 63 | ||
| AK147399.1 | - | 17 - 18 | 3584 | 63 | ||
| AK147501.1 | - | 19 - 20 | 3568 | 63 | ||
| GU734816.2 | - | 19 - 20 | 3128 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp2b2
Genomic Map