Assay Details
| Gene Symbol: Cpne9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170673.3 | NP_733773.2 | 18 - 19 | 1514 | 105 | |
| GenBank mRNA | AK042692.1 | - | 18 - 19 | 1515 | 105 | |
| AK044080.1 | - | 18 - 19 | 1480 | 105 | ||
| AK047571.1 | - | 16 - 17 | 2529 | 105 | ||
| AK137422.1 | - | 18 - 19 | 1515 | 105 | ||
| AK220550.1 | - | 10 - 11 | 2878 | 105 | ||
| BC115728.1 | - | 18 - 19 | 1464 | 40039295 | 105 | |
| BQ550677.1 | - | 3 - 4 | 105 | |||
| BQ550678.1 | - | 2 - 3 | 149 | 105 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cpne9
Genomic Map