| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600009897.mm.1 |
TC0600001258.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600009897.mm.1 |
NM_001113553.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600009897.mm.1 |
NM_001113553.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600009897.mm.1 |
NM_172161.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600009897.mm.1 |
NM_172161.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600009893.mm.1 |
TC0600001258.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600009893.mm.1 |
NM_001113553.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600009893.mm.1 |
NM_001113553.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600009893.mm.1 |
NM_172161.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600009893.mm.1 |
NM_172161.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600009895.mm.1 |
TC0600001258.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600009895.mm.1 |
NM_001113553.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600009895.mm.1 |
NM_001113553.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600009895.mm.1 |
NM_172161.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600009895.mm.1 |
NM_172161.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600009896.mm.1 |
TC0600001258.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600009896.mm.1 |
NM_001113553.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600009896.mm.1 |
NM_001113553.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600009896.mm.1 |
NM_172161.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600009896.mm.1 |
NM_172161.4 |
Refseq |