Assay Details
| Gene Symbol: Il17re | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034029.1 | NP_001029201.1 | 10 - 11 | 1086 | 59 | |
| NM_001034031.1 | NP_001029203.1 | 12 - 13 | 1161 | 59 | ||
| NM_145826.5 | NP_665825.2 | 11 - 12 | 1513 | 59 | ||
| GenBank mRNA | AF458068.1 | - | 11 - 12 | 1163 | 59 | |
| AK046302.1 | - | 11 - 12 | 1516 | 59 | ||
| AK046713.1 | - | 11 - 12 | 1175 | 59 | ||
| AK078876.1 | - | 3 - 4 | 1427 | 59 | ||
| AK169498.1 | - | 11 - 12 | 1341 | 59 | ||
| BC069861.1 | - | 11 - 12 | 1190 | 6311568 | 59 | |
| DQ092339.1 | - | 11 - 12 | 1102 | 59 | ||
| DQ092340.1 | - | 10 - 11 | 1086 | 59 | ||
| DQ092341.1 | - | 12 - 13 | 1161 | 59 | ||
More Information
Set Membership: |
|

Il17re
Genomic Map