Assay Details
| Gene Symbol: Dguok | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013764.2 | NP_038792.2 | 5 - 6 | 783 | 70 | |
| GenBank mRNA | AJ133749.1 | - | 5 - 6 | 708 | 70 | |
| AK035281.1 | - | 5 - 6 | 729 | 70 | ||
| BC012647.1 | - | 5 - 6 | 728 | 70 | ||
| U90524.1 | - | 5 - 6 | 703 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dguok
Genomic Map