Assay Details
Gene Symbol: Nagk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164187.1 | NP_001157659.1 | 2 - 3 | 300 | 74 | |
NM_019542.2 | NP_062415.1 | 2 - 3 | 309 | 74 | ||
GenBank mRNA | AJ242909.1 | - | 2 - 3 | 321 | 74 | |
AJ242911.1 | - | 2 - 3 | 261 | 74 | ||
AK007240.1 | - | 2 - 3 | 304 | 74 | ||
AK155292.1 | - | 2 - 3 | 201 | 74 | ||
BC004689.1 | - | 2 - 3 | 187 | 3501470 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001164187.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
