Assay Details
| Gene Symbol: Nagk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164187.1 | NP_001157659.1 | 6 - 7 | 766 | 68 | |
| NM_019542.2 | NP_062415.1 | 6 - 7 | 775 | 68 | ||
| GenBank mRNA | AJ242909.1 | - | 6 - 7 | 787 | 68 | |
| AJ242911.1 | - | 6 - 7 | 727 | 68 | ||
| AK007240.1 | - | 6 - 7 | 770 | 68 | ||
| AK155292.1 | - | 6 - 7 | 667 | 68 | ||
| BC004689.1 | - | 6 - 7 | 653 | 3501470 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nagk
Genomic Map