Assay Details
| Gene Symbol: Stambp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024239.2 | NP_077201.1 | 3 - 4 | 394 | 94 | |
| GenBank mRNA | AB010123.1 | - | 3 - 4 | 341 | 94 | |
| AK017600.1 | - | 3 - 4 | 349 | 94 | ||
| AK019907.1 | - | 3 - 4 | 344 | 94 | ||
| AK046995.1 | - | 3 - 4 | 346 | 94 | ||
| AK136961.1 | - | 3 - 4 | 945 | 94 | ||
| AK139391.1 | - | 3 - 4 | 369 | 94 | ||
| AK168810.1 | - | 3 - 4 | 367 | 94 | ||
| BC003497.1 | - | 3 - 4 | 337 | 3153829 | 94 | |
| BC006939.1 | - | 3 - 4 | 392 | 2649718 | 94 | |
| BC025111.1 | - | 3 - 4 | 336 | 4194332 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stambp
Genomic Map