Assay Details
Gene Symbol: Bola3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175277.4 | NP_780486.1 | 2 - 3 | 201 | 99 | |
GenBank mRNA | AK019037.1 | - | 1 - 2 | 36 | 99 | |
AK028102.1 | - | 2 - 3 | 202 | 99 | ||
AK131950.1 | - | 210 | 99 | |||
AK160488.1 | - | 2 - 3 | 205 | 99 | ||
BC049748.1 | - | 2 - 3 | 221 | 99 | ||
BC116947.1 | - | 2 - 4 | 40126266 | 99 | ||
BC116949.1 | - | 2 - 4 | 40126268 | 99 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
