Assay Details
| Gene Symbol: Eno2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK003051.1 | - | 2 - 3 | 181 | 95 | |
| AK131652.1 | - | 2 - 3 | 220 | 95 | ||
| AK134571.1 | - | 1 - 2 | 104 | 95 | ||
| AK146655.1 | - | 2 - 3 | 186 | 95 | ||
| AK170169.1 | - | 1 - 2 | 101 | 95 | ||
| BC009018.1 | - | 2 - 3 | 156 | 95 | ||
| BC031739.1 | - | 2 - 3 | 174 | 4527274 | 95 | |
| BY282442.1 | - | 2 - 3 | 337 | 95 | ||
| CA751156.1 | - | 2 - 3 | 218 | 95 | ||
| X52380.1 | - | 2 - 3 | 199 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Eno2
Genomic Map