Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600024112.mm.1 |
TC0600003114.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600024112.mm.1 |
NM_001077705.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600024099.mm.1 |
TC0600003114.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600024099.mm.1 |
NM_001077705.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600024099.mm.1 |
NM_013545.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600024100.mm.1 |
TC0600003114.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600024100.mm.1 |
NM_001077705.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600024100.mm.1 |
NM_013545.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600024110.mm.1 |
TC0600003114.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600024110.mm.1 |
NM_001077705.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600024098.mm.1 |
TC0600003114.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600024098.mm.1 |
NM_001077705.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600024098.mm.1 |
NM_013545.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600024111.mm.1 |
TC0600003114.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600024111.mm.1 |
NM_001077705.2 |
Refseq |