Assay Details
Gene Symbol: Spsb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF403037.1 | - | 1 - 2 | 662 | 62 | |
AI849155.1 | - | 1 - 2 | 62 | |||
AK148177.1 | - | 1 - 2 | 758 | 62 | ||
AK209543.1 | - | 1 - 2 | 109 | 62 | ||
BC002005.1 | - | 2 - 3 | 846 | 3496382 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24817040 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24817041 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24817042 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2127303 | NM_013539.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2127303 | AK148177.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2127303 | BC002005.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2127303 | BC010305.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 463477 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P104869 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2127303 | NM_013539.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2127303 | AK148177.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2127303 | BC002005.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2127303 | BC010305.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817036 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817060 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817049 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817047 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817064 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817057 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817029 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817031 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817053 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817038 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817050 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817062 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817067 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817041 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817033 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817059 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817052 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817055 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817034 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817043 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817069 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817045 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817039 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817035 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P104869 | - | - |
Whole Mouse Genome G4122A | A_51_P410101 | NM_013539.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P410101 | AK148177.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P410101 | BC002005.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P410101 | BC010305.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115986 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P104869 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05471777 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05471765 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05813327 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05471749 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05277165 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05471766 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05471746 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24817042 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24817040 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24817041 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817090 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817143 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817048 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817050 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817088 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817092 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817036 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817086 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817039 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817094 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817099 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817103 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817084 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817052 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817067 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817057 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817144 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817069 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817064 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817053 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817055 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817139 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817045 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817062 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817141 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817034 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817091 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817095 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817033 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817047 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817059 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817093 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817085 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817049 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817041 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24817057 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24817050 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24817066 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24817045 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24817041 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05884145 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05140021 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24817062 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05826121 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24817069 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24817038 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05277165 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24817053 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24817067 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24817052 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24817035 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05846999 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05471757 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05471750 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24817039 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24817028 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24817033 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24817034 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24817059 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24817048 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05472455 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P04998447 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24817055 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10541725 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10541724 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10541723 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10541722 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10541728 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10541727 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1422106_PM_a_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1451585_PM_x_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10541721 | - | - |
Mouse Genome 430 2.0 Array | 1422106_a_at | - | - |
Mouse Genome 430 2.0 Array | 1451585_x_at | - | - |
Mouse Genome 430 2.0 Array | 1422106_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1451585_PM_x_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14794_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14794_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0600011548.mm.1 | TC0600001432.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600011548.mm.1 | NM_013539.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600011547.mm.1 | TC0600001432.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600011547.mm.1 | NM_013539.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600011545.mm.1 | TC0600001432.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600011545.mm.1 | NM_013539.2 | Refseq |
Mouse Expression Set 430 | 1422106_a_at | - | - |
Mouse Expression Set 430 | 1422106_PM_a_at | - | - |
Mouse Expression Set 430 | 1451585_x_at | - | - |
Mouse Expression Set 430 | 1451585_PM_x_at | - | - |
Murine Genome U74v2 Set | 103993_at | - | - |
Murine Genome U74v2 Set | 103993_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1422106_PM_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1451585_x_at | - | - |
Mouse Genome 430A 2.0 Array | 1422106_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1451585_PM_x_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1422106_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1451585_x_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1422106_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1451585_PM_x_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 463844 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 840717 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 840717 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000840717 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1254644 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1254644 | - | - |
mouse-6 v1.1 (Illumina) | scl29517.3_59-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 840717 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl29517.3_59-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 840717 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1254644 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1254644 | - | - |
