Assay Details
| Gene Symbol: Tmem57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025382.6 | NP_079658.2 | 3 - 4 | 572 | 74 | |
| GenBank mRNA | AK038972.1 | - | 4 - 5 | 734 | 74 | |
| AY364165.1 | - | 3 - 4 | 357 | 74 | ||
| AY846873.1 | - | 3 - 4 | 573 | 74 | ||
| AY846874.1 | - | 3 - 4 | 573 | 74 | ||
| BC019186.1 | - | 3 - 4 | 541 | 74 | ||
| BC042733.2 | - | 3 - 4 | 512 | 74 | ||
| BC044802.1 | - | 3 - 4 | 512 | 74 | ||
| BC054355.1 | - | 3 - 4 | 376 | 74 | ||
| BC054769.1 | - | 3 - 4 | 465 | 6415086 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem57
Genomic Map