Assay Details
| Gene Symbol: Tmem57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025382.6 | NP_079658.2 | 7 - 8 | 1534 | 84 | |
| GenBank mRNA | AY364165.1 | - | 7 - 8 | 1319 | 84 | |
| AY846873.1 | - | 7 - 8 | 1535 | 84 | ||
| AY846874.1 | - | 7 - 8 | 1535 | 84 | ||
| BC019186.1 | - | 7 - 8 | 1503 | 84 | ||
| BC043306.1 | - | 4 - 5 | 966 | 84 | ||
| BC054355.1 | - | 8 - 9 | 1447 | 84 | ||
| BC054769.1 | - | 7 - 8 | 1427 | 6415086 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem57
Genomic Map