Assay Details
Gene Symbol: Cep85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144527.3 | NP_653110.3 | 6 - 7 | 1308 | 100 | |
GenBank mRNA | AK030742.1 | - | 6 - 7 | 1273 | 100 | |
AK085545.1 | - | 6 - 7 | 1255 | 100 | ||
AK141103.1 | - | 6 - 7 | 1298 | 100 | ||
BC008221.1 | - | 6 - 7 | 1260 | 100 | ||
BC031729.1 | - | 6 - 7 | 1198 | 3992639 | 100 | |
BC114391.1 | - | 2 - 3 | 204 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
