Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400011887.mm.1 |
NM_001164745.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011887.mm.1 |
NM_001164745.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011887.mm.1 |
NM_008974.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011887.mm.1 |
NM_008974.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011887.mm.1 |
TC0400001530.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400011888.mm.1 |
NM_001164745.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011888.mm.1 |
NM_001164745.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011888.mm.1 |
NM_008974.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011888.mm.1 |
NM_008974.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011888.mm.1 |
TC0400001530.mm.1 |
MTA TC |