Assay Details
| Gene Symbol: Armc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034858.3 | NP_001030030.2 | 15 - 16 | 2345 | 72 | |
| GenBank mRNA | AK141039.1 | - | 9 - 10 | 2677 | 72 | |
| BC172168.1 | - | 10 - 11 | 2087 | 72 | ||
More Information
Set Membership: |
|

Armc2
Genomic Map