| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1000014706.mm.1 |
NM_001034858.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000014706.mm.1 |
NM_001034858.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000014706.mm.1 |
TC1000002057.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000014701.mm.1 |
NM_001034858.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000014701.mm.1 |
NM_001034858.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000014701.mm.1 |
TC1000002057.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000014704.mm.1 |
NM_001034858.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000014704.mm.1 |
NM_001034858.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000014704.mm.1 |
TC1000002057.mm.1 |
MTA TC |