Assay Details
| Gene Symbol: Armc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034858.3 | NP_001030030.2 | 8 - 9 | 1224 | 77 | |
| GenBank mRNA | AK131686.1 | - | 4 - 5 | 419 | 77 | |
| AK141039.1 | - | 2 - 3 | 1556 | 77 | ||
| BC172168.1 | - | 3 - 4 | 966 | 77 | ||
More Information
Set Membership: |
|

Armc2
Genomic Map