Assay Details
| Gene Symbol: Armc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034858.3 | NP_001030030.2 | 11 - 12 | 1699 | 67 | |
| GenBank mRNA | AK141039.1 | - | 5 - 6 | 2031 | 67 | |
| BC172168.1 | - | 6 - 7 | 1441 | 67 | ||
More Information
Set Membership: |
|

Armc2
Genomic Map