Assay Details
| Gene Symbol: Armc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034858.3 | NP_001030030.2 | 12 - 13 | 1808 | 87 | |
| GenBank mRNA | AK131686.1 | - | 8 - 9 | 1005 | 87 | |
| AK141039.1 | - | 6 - 7 | 2140 | 87 | ||
| BC172168.1 | - | 7 - 8 | 1550 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Armc2
Genomic Map